Gene Transcripts

Gene Symbol
Nxt2
Entrez Gene ID
237082
Gene Name
nuclear transport factor 2-like export factor 2
Gene Aliases
6330587F24Rik, AI787442, P15-2
Chromosome Location
Chr. X: 142226770 - 142239700 on Build GRCm38
UniGene ID
Mm.41899
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161430.2 NP_001154902.1 4 291
NM_001290532.1 NP_001277461.1 4 376
NM_001290533.1 NP_001277462.1 3 382
NM_172782.4 NP_766370.2 3 328
GenBank mRNA AK032102.1 - 3 299
AK144349.1 - 3 312
BB851047.1 - 3 330
BB857403.1 - 4 292
BC064727.1 - 3 303
BC068166.1 - 3 281 30361776
BC094570.1 - 3 245 30361291
BC118503.1 - 3 256 40106658
CA482101.1 - 4 327
CJ148018.1 - 4 377

View Full Product Details