Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081656.2 | NP_001075125.1 | 5 | 5474 | |
GenBank mRNA | AK030658.1 | - | 1 | 773 | |
AK053917.1 | - | 1 | 2105 | ||
AK082655.1 | - | 1 | 2156 | ||
BC150855.1 | - | 1 | 1895 | ||
BC150866.1 | - | 1 | 1897 |