Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004860.3 | NP_004851.2 | 1, 2 | 454 | |
GenBank mRNA | AK313836.1 | - | 1, 2 | 139 | |
BC020090.1 | - | 1, 2 | 408 | 4385430 | |
BC051907.1 | - | 1, 2 | 285 | 6149976 | |
BC067272.1 | - | 1, 2 | 399 | 6204341 | |
BT009817.1 | - | 1, 2 | 89 | ||
U31501.1 | - | 1, 2 | 316 |