Gene Transcripts

Gene Symbol
Rtn4rl1
Entrez Gene ID
237847
Gene Name
reticulon 4 receptor-like 1
Gene Aliases
Ngr3, Ngrh2, Ngrl2
Chromosome Location
Chr. 11: 75193993 - 75267769 on Build GRCm38
UniGene ID
Mm.82661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177708.5 NP_808376.1 2 2295
GenBank mRNA AK033286.1 - 2 2293
AK140456.1 - 2 2296
BC030471.1 - 2 2499 5368560

View Full Product Details