Gene Transcripts
- Gene Symbol
- GMFG
- Entrez Gene ID
- 9535
- Gene Name
- glia maturation factor gamma
- Gene Aliases
- GMF-GAMMA
- Chromosome Location
- Chr. 19: 39328357 - 39336086 on Build GRCh38
- UniGene ID
- Hs.5210
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301008.1 |
NP_001287937.1
|
3, 4 |
221 |
|
NM_004877.3 |
NP_004868.1
|
4, 5 |
318 |
|
GenBank mRNA |
AB001993.1 |
- |
3, 4 |
218 |
|
AF038956.1 |
- |
3, 4 |
212 |
|
AV714415.1 |
- |
3, 4 |
152 |
|
BC022207.1 |
- |
4, 5 |
238 |
|
BC032819.2 |
- |
3, 4 |
205 |
|
BC080180.1 |
- |
4, 5 |
238 |
5419511 |
BC093799.1 |
- |
4, 5 |
271 |
7939644 |
BC101818.1 |
- |
4, 5 |
271 |
8069324 |
BC143543.1 |
- |
4, 5 |
271 |
9052054 |
BC143546.1 |
- |
4, 5 |
270 |
9052057 |
BC143548.1 |
- |
4, 5 |
269 |
9052059 |
BF974742.1 |
- |
4, 5 |
319 |
|
CR456967.1 |
- |
3, 4 |
208 |
|
CR542114.1 |
- |
3, 4 |
208 |
|
View Full Product Details