Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010686.3 | NP_034816.1 | 8 | 2321 | |
GenBank mRNA | AK031802.1 | - | 8 | 2333 | |
AK089508.1 | - | 9 | 2272 | ||
AK089535.1 | - | 9 | 2264 | ||
AK150299.1 | - | 9 | 2299 | ||
AK152918.1 | - | 9 | 1936 | ||
AK153877.1 | - | 8 | 2335 | ||
AK155353.1 | - | 3 | 1778 | ||
AK155744.1 | - | 9 | 2153 | ||
AK163146.1 | - | 8 | 2336 | ||
AK170690.1 | - | 10 | 2128 | ||
AK170867.1 | - | 9 | 2015 | ||
AK171053.1 | - | 9 | 2272 | ||
AK171057.1 | - | 8 | 2372 | ||
AK171116.1 | - | 9 | 2264 | ||
AK171193.1 | - | 9 | 2264 | ||
AK171213.1 | - | 8 | 2336 | ||
AK171236.1 | - | 9 | 2047 | ||
AK171244.1 | - | 8 | 2336 | ||
AK171250.1 | - | 9 | 2105 | ||
AK171269.1 | - | 8 | 2373 | ||
AK171270.1 | - | 8 | 2339 | ||
AK171274.1 | - | 8 | 2329 | ||
AK171280.1 | - | 8 | 2369 | ||
AK171285.1 | - | 8 | 2373 | ||
AK171481.1 | - | 8 | 2409 | ||
AK171547.1 | - | 9 | 2233 | ||
AK171562.1 | - | 9 | 2258 | ||
AK171571.1 | - | 9 | 2284 | ||
AK171575.1 | - | 9 | 2302 | ||
AK172306.1 | - | 9 | 1980 | ||
AK172486.1 | - | 8 | 2371 | ||
AK172707.1 | - | 9 | 2048 | ||
AK172717.1 | - | 6 | 1970 | ||
BC020993.1 | - | 8 | 2403 | 4037686 | |
BC055057.1 | - | 8 | 2390 | 6529760 | |
U29539.1 | - | 8 | 2377 |