Gene Transcripts

Gene Symbol
Olfr49
Entrez Gene ID
18348
Gene Name
olfactory receptor 49
Gene Aliases
IC6, MOR1181
Chromosome Location
Chr. 14: 54281896 - 54282925 on Build GRCm38
UniGene ID
Mm.436623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010991.2 NP_035121.1 1 360
GenBank mRNA BC145891.1 - 1 360
BC145893.1 - 1 360 40131196

View Full Product Details