Gene Transcripts

Gene Symbol
Cdx4
Entrez Gene ID
12592
Gene Name
caudal type homeobox 4
Gene Aliases
Cdx-3, Cdx-4, Cdx3
Chromosome Location
Chr. X: 103321398 - 103330224 on Build GRCm38
UniGene ID
Mm.4353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007674.3 NP_031700.1 3 1001
GenBank mRNA AK141681.1 - 3 415
BC119158.1 - 3 989 8733907
L08061.1 - 3 1004

View Full Product Details