Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007684.3 | NP_031710.1 | 4 | 413 | |
GenBank mRNA | AK003473.1 | - | 4 | 429 | |
AK004532.1 | - | 4 | 425 | ||
AK006289.1 | - | 4 | 428 | ||
AK006891.1 | - | 4 | 413 | ||
AK011821.1 | - | 4 | 406 | ||
AK017505.1 | - | 4 | 453 | ||
BC002162.1 | - | 4 | 447 | 3485864 | |
BC054097.1 | - | 4 | 449 | 30019209 | |
Y12474.1 | - | 4 | 364 |