Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009755.3 | NP_033885.2 | 12 | 1842 | |
GenBank mRNA | AK004995.2 | - | 12 | 1477 | |
AK147534.1 | - | 10 | 1245 | ||
AK158218.1 | - | 12 | 1835 | ||
AK179671.1 | - | 2 | 178 | ||
AK220295.1 | - | 12 | 1853 | ||
BC003925.1 | - | 2 | 208 | ||
BC053923.1 | - | 12 | 1769 | ||
BC063079.1 | - | 8 | 972 | ||
BC066062.1 | - | 12 | 1770 | 6849066 | |
L24755.1 | - | 12 | 1619 |