Gene Transcripts

Gene Symbol
Tas2r105
Entrez Gene ID
57252
Gene Name
taste receptor, type 2, member 105
Gene Aliases
T2R05, T2R9, T2r5, Tas2r5, mGR05, mt2r5
Chromosome Location
Chr. 6: 131686561 - 131687463 on Build GRCm38
UniGene ID
Mm.377131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020501.1 NP_065247.1 1 458
GenBank mRNA BC125515.1 - 1 490
BC125541.1 - 1 490 40130056

View Full Product Details