Gene Transcripts

Gene Symbol
Emid1
Entrez Gene ID
140703
Gene Name
EMI domain containing 1
Gene Aliases
AW122071, CO-5, Emu1
Chromosome Location
Chr. 11: 5106265 - 5152298 on Build GRCm38
UniGene ID
Mm.840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080595.2 NP_542162.1 15 1467
GenBank mRNA AJ416093.1 - 15 1467
AK153702.1 - 17 1645
BC012248.1 - 6 569
BC021849.1 - 5 471

View Full Product Details