Gene Transcripts

Gene Symbol
Sirt5
Entrez Gene ID
68346
Gene Name
sirtuin 5
Gene Aliases
0610012J09Rik, 1500032M05Rik, AV001953
Chromosome Location
Chr. 13: 43365439 - 43395203 on Build GRCm38
UniGene ID
Mm.35325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178848.3 NP_849179.1 8 976
GenBank mRNA AK002609.1 - 9 1005
BC031770.1 - 8 976 4014119

View Full Product Details