Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008617.2 | NP_032643.2 | 8 | 906 | |
GenBank mRNA | AK002305.1 | - | 8 | 923 | |
AK135162.1 | - | 8 | 919 | ||
AK160553.1 | - | 8 | 889 | ||
AK167809.1 | - | 8 | 889 | ||
BC023482.1 | - | 8 | 906 | 5251192 | |
DQ402950.1 | - | 7 | 756 | ||
M16229.1 | - | 8 | 865 |