Gene Transcripts

Gene Symbol
Tex2
Entrez Gene ID
21763
Gene Name
testis expressed gene 2
Gene Aliases
4930568E07Rik, AI553404, Def-5, Taz4
Chromosome Location
Chr. 11: 106502139 - 106613418 on Build GRCm38
UniGene ID
Mm.102407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198292.3 NP_938034.2 2 325
GenBank mRNA AK029537.1 - 2 332
AK129434.1 - 2 325
AK168397.1 - 2 333
AK212434.1 - 1 105

View Full Product Details