Gene Transcripts

Gene Symbol
ARIH1
Entrez Gene ID
25820
Gene Name
ariadne RBR E3 ubiquitin protein ligase 1
Gene Aliases
ARI, HARI, HHARI, UBCH7BP
Chromosome Location
Chr. 15: 72474282 - 72586563 on Build GRCh38
UniGene ID
Hs.268787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005744.3 NP_005735.2 13 1879
XM_011521438.2 XP_011519740.1 12 1855
XM_011521439.2 XP_011519741.1 14 1393
GenBank mRNA AB014774.1 - 4 439
AB209432.1 - 14 1508
AF072832.1 - 13 1565
AJ009771.1 - 6 675
AJ130976.1 - 13 1283
AJ243190.1 - 13 1879
AK312715.1 - 13 1878
BC051877.1 - 13 1814 5742498
BX641117.1 - 1 429

View Full Product Details