Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027905.2 | NP_082181.2 | 4 | 974 | |
GenBank mRNA | AK005048.1 | - | 4 | 974 | |
AK054432.1 | - | 4 | 936 | ||
AK161059.1 | - | 4 | 925 | ||
BC117817.1 | - | 5 | 489 | ||
BC117818.1 | - | 5 | 522 |