Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153573.1 | NP_705801.1 | 4 | 753 | |
GenBank mRNA | AK030620.1 | - | 4 | 804 | |
AK081692.1 | - | 4 | 780 | ||
AK163384.1 | - | 899 | |||
BC024846.1 | - | 1180 | |||
BC029109.1 | - | 4 | 753 | 3985764 | |
BC034313.1 | - | 5 | 868 | 5365046 |