Gene Transcripts

Gene Symbol
Adap1
Entrez Gene ID
231821
Gene Name
ArfGAP with dual PH domains 1
Gene Aliases
4930431P11Rik, Centa1, GC1L, p42IP4
Chromosome Location
Chr. 5: 139271876 - 139325464 on Build GRCm38
UniGene ID
Mm.297819
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172723.4 NP_766311.2 11 2164
GenBank mRNA AK076768.1 - 11 1922
BC056934.1 - 10 1890
BC079634.1 - 10 1958

View Full Product Details