Gene Transcripts

Gene Symbol
Crybg3
Entrez Gene ID
224273
Gene Name
beta-gamma crystallin domain containing 3
Gene Aliases
5031404N19, AU022790
Chromosome Location
Chr. 16: 59490775 - 59601399 on Build GRCm38
UniGene ID
Mm.246424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174848.3 NP_777273.2 1 1277
GenBank mRNA BC043118.1 - 1 1277

View Full Product Details