Gene Transcripts

Gene Symbol
Onecut2
Entrez Gene ID
225631
Gene Name
one cut domain, family member 2
Gene Aliases
C730009D12, OC-2, Oc2
Chromosome Location
Chr. 18: 64340364 - 64398488 on Build GRCm38
UniGene ID
Mm.234723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194268.2 NP_919244.2 1 672
GenBank mRNA AY242995.1 - 1 672
BC100750.1 - 2 169
BC100755.1 - 2 169
BC103668.1 - 2 505 40046651

View Full Product Details