Gene Transcripts

Gene Symbol
Art4
Entrez Gene ID
109978
Gene Name
ADP-ribosyltransferase 4
Gene Aliases
4432404K01Rik, ARTC4, DO, DOK1
Chromosome Location
Chr. 6: 136848451 - 136857600 on Build GRCm38
UniGene ID
Mm.31840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026639.2 NP_080915.1 3 1449
GenBank mRNA AK004744.1 - 3 1403
AK019467.1 - 3 1450
BC046306.1 - 3 1370 6306234

View Full Product Details