Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172467.3 | NP_766055.1 | 4 | 926 | |
GenBank mRNA | AK045287.1 | - | 4 | 927 | |
AK051064.1 | - | 4 | 928 | ||
AK051832.1 | - | 2 | 314 | ||
AK084475.1 | - | 4 | 858 | ||
BC138180.1 | - | 4 | 1018 | 8861199 | |
BC138181.1 | - | 4 | 1018 | 8861200 |