Gene Transcripts
- Gene Symbol
- Usp8
- Entrez Gene ID
- 84092
- Gene Name
- ubiquitin specific peptidase 8
- Gene Aliases
- AI574262, AW557536, Ubpy, mKIAA0055
- Chromosome Location
- Chr. 2: 126707202 - 126759314 on Build GRCm38
- UniGene ID
- Mm.272629
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252580.1 |
NP_001239509.1
|
4, 5 |
590 |
|
NM_019729.3 |
NP_062703.2
|
4, 5 |
557 |
|
GenBank mRNA |
AB045709.1 |
- |
3, 4 |
331 |
|
AF057146.1 |
- |
3, 4 |
348 |
|
AK122195.1 |
- |
4, 5 |
468 |
|
AK129003.1 |
- |
4, 5 |
558 |
|
AK134058.1 |
- |
4, 5 |
512 |
|
AK142881.1 |
- |
1, 2 |
55 |
|
AK163469.1 |
- |
4, 5 |
490 |
|
AK166540.1 |
- |
4, 5 |
558 |
|
BC038123.1 |
- |
4, 5 |
542 |
|
BC050947.1 |
- |
4, 5 |
522 |
6825981 |
BC061465.1 |
- |
4, 5 |
522 |
6825981 |
BC066126.1 |
- |
4, 5 |
520 |
30301665 |
View Full Product Details