Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190456.1 | NP_001177385.1 | 3 | 292 | |
NM_001190457.1 | NP_001177386.1 | 3 | 571 | ||
NM_001324014.1 | NP_001310943.1 | 4 | 503 | ||
NM_001324015.1 | NP_001310944.1 | 3 | 304 | ||
NM_006091.4 | NP_006082.3 | 3 | 526 | ||
GenBank mRNA | AB010098.1 | - | 3 | 259 | |
AB023142.1 | - | 2 | 230 | ||
AK289678.1 | - | 3 | 571 | ||
BC026335.1 | - | 3 | 242 | 4792415 | |
BX461395.2 | - | 3 | 525 | ||
DA128408.1 | - | 3 | 292 | ||
DA182763.1 | - | 3 | 261 | ||
DA219458.1 | - | 3 | 253 | ||
DA291058.1 | - | 3 | 253 |