Gene Transcripts

Gene Symbol
Mrpl27
Entrez Gene ID
94064
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
A630055F16Rik, D11Moh47, D18Ertd643e
Chromosome Location
Chr. 11: 94653770 - 94660087 on Build GRCm38
UniGene ID
Mm.34951
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053161.2 NP_444391.1 4 318
GenBank mRNA AB049648.1 - 4 313
AK143575.1 - 4 347
BC030074.1 - 4 318 1514322
BC096568.1 - 4 325

View Full Product Details