Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080560.3 | NP_542127.1 | 4 | 1449 | |
GenBank mRNA | AK005302.1 | - | 4 | 1467 | |
AK150930.1 | - | 1 | 390 | ||
AK161968.1 | - | 4 | 1465 | ||
BC067069.1 | - | 4 | 1450 | 30550024 | |
BC132630.1 | - | 4 | 1424 | 40130907 | |
BC138211.1 | - | 4 | 1424 | 8861230 |