Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178936.3 | NP_849267.1 | 4 | 518 | |
GenBank mRNA | AK016295.1 | - | 6 | 772 | |
AK147601.1 | - | 4 | 566 | ||
AK149544.1 | - | 4 | 543 | ||
AK163285.1 | - | 4 | 556 | ||
BC040365.1 | - | 4 | 518 | 4223855 |