Gene Transcripts

Gene Symbol
C1orf61
Entrez Gene ID
10485
Gene Name
chromosome 1 open reading frame 61
Gene Aliases
CROC4
Chromosome Location
Chr. 1: 156404252 - 156456649 on Build GRCh38
UniGene ID
Hs.380027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006365.2 NP_006356.1 5, 6 584
NR_135264.1 4, 5 488
NR_135268.1 5, 6 818
XM_011509070.1 XP_011507372.1 4, 5 897
XM_011509071.2 XP_011507373.1 4, 5 1684
XM_011509072.1 XP_011507374.1 4, 5 507
XM_011509073.2 XP_011507375.1 3, 4 671
XM_011509074.1 XP_011507376.1 3, 4 452
XM_017000062.1 XP_016855551.1 8, 9 1065
XM_017000063.1 XP_016855552.1 6, 7 689
GenBank mRNA BI752228.1 - 5, 6 807
DB534652.2 - 2, 3 263
HY158402.1 - 4, 5 366
U49857.1 - 5, 6 428

View Full Product Details