Gene Transcripts

Gene Symbol
Spag17
Entrez Gene ID
74362
Gene Name
sperm associated antigen 17
Gene Aliases
4931427F14Rik, PF6, Spag17-ps
Chromosome Location
Chr. 3: 99885417 - 100143322 on Build GRCm38
UniGene ID
Mm.55847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028892.4 NP_083168.3 18 2528
GenBank mRNA AK029691.1 - 18 2529
AY555275.1 - 18 2514
AY792594.1 - 18 2514

View Full Product Details