Gene Transcripts

Gene Symbol
Slc9b1
Entrez Gene ID
74446
Gene Name
solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1
Gene Aliases
1700094G20Rik, 4933424B12Rik, 4933425K02Rik, Nhedc1, mtsNHE
Chromosome Location
Chr. 3: 135347994 - 135397827 on Build GRCm38
UniGene ID
Mm.85000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028946.3 NP_083222.2 10 1694
GenBank mRNA AK016917.1 - 10 1613
AK029525.1 - 10 1695
EU846100.1 - 9 1225

View Full Product Details