Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145824.4 | NP_665823.2 | 11 | 1554 | |
GenBank mRNA | AK173177.1 | - | 11 | 1367 | |
AY337314.1 | - | 11 | 1555 | ||
BC024698.1 | - | 11 | 1399 | 3667000 | |
EU281316.1 | - | 11 | 1415 |