Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_182994.2 | NP_892039.1 | 5 | 599 | |
GenBank mRNA | AK147525.1 | - | 5 | 599 | |
BC048170.1 | - | 5 | 599 | 6821976 | |
BC059931.1 | - | 5 | 599 | ||
BC100307.1 | - | 5 | 596 | 30843038 |