Gene Transcripts

Gene Symbol
Dhrs13
Entrez Gene ID
70451
Gene Name
dehydrogenase/reductase (SDR family) member 13
Gene Aliases
2610209N15Rik
Chromosome Location
Chr. 11: 78032270 - 78037866 on Build GRCm38
UniGene ID
Mm.390342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183286.2 NP_899109.2 3 389
GenBank mRNA AK011939.1 - 3 364
AK046465.1 - 2 436
BB637905.1 - 3 389
BC115881.1 - 2 234 40088339

View Full Product Details