Gene Transcripts

Gene Symbol
Fads3
Entrez Gene ID
60527
Gene Name
fatty acid desaturase 3
Gene Aliases
AI464531
Chromosome Location
Chr. 19: 10040855 - 10059671 on Build GRCm38
UniGene ID
Mm.253875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021890.3 NP_068690.3 12 3220
GenBank mRNA AK029318.1 - 12 3079
AK090042.1 - 12 3206
AK154534.1 - 12 3097
AK162624.1 - 1 1256

View Full Product Details