Gene Transcripts

Gene Symbol
Cml1
Entrez Gene ID
66116
Gene Name
camello-like 1
Gene Aliases
1110002I11Rik, AI266962
Chromosome Location
Chr. 6: 85910154 - 85915677 on Build GRCm38
UniGene ID
Mm.13036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023160.2 NP_075649.2 2 969
GenBank mRNA AF163314.1 - 2 967
AK002975.1 - 1 323
AK003301.1 - 2 919
BC010796.1 - 2 923 4223199

View Full Product Details