Gene Transcripts

Gene Symbol
Bfar
Entrez Gene ID
67118
Gene Name
bifunctional apoptosis regulator
Gene Aliases
3010001A07Rik, AI666707, AW107665, Bar, Rnf47
Chromosome Location
Chr. 16: 13671858 - 13703612 on Build GRCm38
UniGene ID
Mm.223689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177552.1 NP_001171023.1 3 602
NM_025976.5 NP_080252.1 5 977
GenBank mRNA AK013874.1 - 5 842
AK028592.1 - 3 441
AK041839.1 - 4 716
BC027221.1 - 5 818 4196466

View Full Product Details