Gene Transcripts
- Gene Symbol
- Tuba1a
- Entrez Gene ID
- 22142
- Gene Name
- tubulin, alpha 1A
- Gene Aliases
- Tuba-1, Tuba1
- Chromosome Location
- Chr. 15: 98949847 - 98953501 on Build GRCm38
- UniGene ID
- Mm.405359
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M13445.1 |
- |
4 |
734 |
|
View Full Product Details