Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011663.3 | NP_035793.1 | 1 | 2215 | |
GenBank mRNA | AK030552.1 | - | 1 | 1005 | |
BC132555.1 | - | 2 | 1273 | 40130832 | |
BC138594.1 | - | 1 | 1283 | 8861614 | |
D17407.1 | - | 2 | 2160 | ||
S69507.1 | - | 1 | 2149 |