Gene Transcripts

Gene Symbol
Xcr1
Entrez Gene ID
23832
Gene Name
chemokine (C motif) receptor 1
Gene Aliases
Ccxcr1, Gpr5, mXcr1
Chromosome Location
Chr. 9: 123852315 - 123862029 on Build GRCm38
UniGene ID
Mm.390241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011798.4 NP_035928.2 2 4320
GenBank mRNA AK040878.1 - 2 4321

View Full Product Details