Gene Transcripts

Gene Symbol
Cenph
Entrez Gene ID
26886
Gene Name
centromere protein H
Gene Aliases
1700021I11Rik, 2410018A12Rik, 2610042E16Rik, 2810046K12Rik, AU044255, CENP-H, ENP
Chromosome Location
Chr. 13: 100759686 - 100775899 on Build GRCm38
UniGene ID
Mm.273502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021886.1 NP_068686.1 2 246
GenBank mRNA AB017634.1 - 2 246
AK011264.1 - 2 231
AK011738.1 - 2 231
BC025084.1 - 2 197 3501244

View Full Product Details