Gene Transcripts

Gene Symbol
Itpka
Entrez Gene ID
81677
Gene Name
inositol-trisphosphate 3-kinase A
Gene Aliases
Chromosome Location
Chr. 3: 111545007 - 111553605 on Build Rnor_6.0
UniGene ID
Rn.9877
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031045.2 NP_112307.2 2 572
GenBank mRNA M29787.1 - 2 572
X56917.1 - 2 563

View Full Product Details