Gene Transcripts
- Gene Symbol
- THBS3
- Entrez Gene ID
- 7059
- Gene Name
- thrombospondin 3
- Gene Aliases
- TSP3
- Chromosome Location
- Chr. 1: 155195588 - 155209180 on Build GRCh38
- UniGene ID
- Hs.169875
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252607.1 |
NP_001239536.1
|
1, 2 |
191 |
|
NM_001252608.1 |
NP_001239537.1
|
1, 2 |
191 |
|
NM_007112.4 |
NP_009043.1
|
1, 2 |
191 |
|
NR_045553.1 |
|
1, 2 |
191 |
|
XM_011509929.1 |
XP_011508231.1
|
1, 2 |
146 |
|
XM_011509934.1 |
XP_011508236.1
|
1, 2 |
126 |
|
XM_011509936.1 |
XP_011508238.1
|
1, 2 |
126 |
|
XM_017002206.1 |
XP_016857695.1
|
1, 2 |
126 |
|
GenBank mRNA |
AK223278.1 |
- |
1, 2 |
105 |
|
AK298592.1 |
- |
1, 2 |
127 |
|
AK298803.1 |
- |
1, 2 |
109 |
|
AK314337.1 |
- |
1, 2 |
142 |
|
BC113846.1 |
- |
1, 2 |
109 |
40008346 |
BC113847.1 |
- |
1, 2 |
109 |
40008343 |
BX490216.1 |
- |
1, 2 |
110 |
|
L38969.1 |
- |
1, 2 |
107 |
|
View Full Product Details