Gene Transcripts

Gene Symbol
Gpr56
Entrez Gene ID
260326
Gene Name
G protein-coupled receptor 56
Gene Aliases
Chromosome Location
Chr. 19: 10423534 - 10460716 on Build Rnor_6.0
UniGene ID
Rn.1677
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152242.2 NP_689448.2 3 546
XM_006255066.2 XP_006255128.1 3 1598
XM_008772264.1 XP_008770486.1 3 748
XM_008772265.1 XP_008770487.1 4 1012
XM_008772266.1 XP_008770488.1 3 2258
XM_008772267.1 XP_008770489.1 3 2521
XM_008772268.1 XP_008770490.1 4 845
XM_008772269.1 XP_008770491.1 3 1627
XM_008772270.1 XP_008770492.1 3 2103
GenBank mRNA AF529886.1 - 3 547
CO557428.1 - 3 560

View Full Product Details