Gene Transcripts

Gene Symbol
PWP1
Entrez Gene ID
11137
Gene Name
PWP1 homolog, endonuclein
Gene Aliases
IEF-SSP-9502
Chromosome Location
Chr. 12: 107685732 - 107713162 on Build GRCh38
UniGene ID
Hs.506652
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317962.1 NP_001304891.1 3 171
NM_001317963.1 NP_001304892.1 3 315
NM_007062.2 NP_008993.1 3 315
GenBank mRNA AK223162.1 - 3 239
AK290681.1 - 3 156
AK296252.1 - 3 172
AK297961.1 - 3, 4 267
BC000067.2 - 3 198
BC001652.2 - 3 234 2924215
BC010921.1 - 3 249
BC032127.2 - 3 237
BC040135.1 - 3 213
BC046144.1 - 3 229
DC380488.1 - 3 315
L07758.1 - 3 234

View Full Product Details