Gene Transcripts

Gene Symbol
Fads3
Entrez Gene ID
286922
Gene Name
fatty acid desaturase 3
Gene Aliases
Chromosome Location
Chr. 1: 226091774 - 226108332 on Build Rnor_6.0
UniGene ID
Rn.35262
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173137.1 NP_775160.1 4, 5 726
GenBank mRNA AJ494720.1 - 4, 5 726
HQ901598.1 - 4, 5 650

View Full Product Details