Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007124.2 | NP_009055.2 | 3 | 314 | |
XM_005267127.4 | XP_005267184.1 | 3 | 649 | ||
XM_005267130.2 | XP_005267187.1 | 4 | 532 | ||
XM_005267133.2 | XP_005267190.1 | 3 | 1332 | ||
XM_011536101.2 | XP_011534403.1 | 5 | 993 | ||
XM_011536102.2 | XP_011534404.1 | 5 | 534 | ||
XM_011536106.2 | XP_011534408.1 | 4 | 1181 | ||
XM_011536107.2 | XP_011534409.1 | 4 | 557 | ||
XM_017011243.1 | XP_016866732.1 | 5 | 534 | ||
XM_017011244.1 | XP_016866733.1 | 5 | 534 | ||
XM_017011245.1 | XP_016866734.1 | 5 | 534 | ||
GenBank mRNA | AK292506.1 | - | 4 | 532 | |
DB079517.1 | - | 4 | 532 | ||
X69086.1 | - | 3 | 222 |