Gene Transcripts

Gene Symbol
Prl8a7
Entrez Gene ID
64368
Gene Name
prolactin family 8, subfamily a, member 7
Gene Aliases
Prlpd
Chromosome Location
Chr. 17: 39509516 - 39515418 on Build Rnor_6.0
UniGene ID
Rn.202970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022537.1 NP_071982.1 2 249
XM_006253909.1 XP_006253971.1 2 223
GenBank mRNA AB000107.1 - 2 249
BC098917.1 - 2 229 7445772

View Full Product Details