Gene Transcripts

Gene Symbol
Rpl14
Entrez Gene ID
65043
Gene Name
ribosomal protein L14
Gene Aliases
Chromosome Location
Chr. 8: 129240528 - 129243400 on Build Rnor_6.0
UniGene ID
Rn.108039
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022949.2 NP_075238.2 4 256
GenBank mRNA BC168728.1 - 4 256
X94242.1 - 3 253

View Full Product Details