Gene Transcripts

Gene Symbol
Shank3
Entrez Gene ID
59312
Gene Name
SH3 and multiple ankyrin repeat domains 3
Gene Aliases
Prosap2
Chromosome Location
Chr. 7: 130474278 - 130534679 on Build Rnor_6.0
UniGene ID
Rn.42876
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021676.1 NP_067708.1 21 4750
GenBank mRNA AF133301.1 - 21 4750
AJ133120.1 - 21 4751

View Full Product Details